siRNA Details
| Gene Symbol: Rnf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146025.1 | NP_001139497.1 | 7 | 1192 | ||
| NM_001146026.1 | NP_001139498.1 | 7 | 1117 | |||
| NM_001146027.1 | NP_001139499.1 | 7 | 983 | |||
| NM_134064.2 | NP_598825.2 | 7 | 1125 | |||
| GenBank mRNA | AK147349.1 | - | 7 | 1118 | ||
| AK160090.1 | - | 7 | 976 | |||
| AK160115.1 | - | 7 | 976 | |||
| AK161651.1 | - | 7 | 1105 | |||
| AK171943.1 | - | 7 | 974 | |||
| BC017630.1 | - | 7 | 918 | 4507289 | ||
| BC035548.1 | - | 7 | 940 | 4221284 | ||

Rnf44
Genomic Map