siRNA Details
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142152.1 | - | 5, 6 | 665 | ||
AK154645.1 | - | 3, 4 | 2266 | |||
AK163238.1 | - | 3, 4 | 2250 | |||
BC031484.1 | - | 5, 6 | 660 | 2654265 | ||
BC047372.1 | - | 5, 6 | 644 | |||
BC057597.1 | - | 3, 4 | 2228 | |||
BC089379.1 | - | 5, 6 | 618 |
