siRNA Details
| Gene Symbol: AW209491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104646.1 | NP_001098116.1 | 2 | 1285 | ||
| NM_134067.4 | NP_598828.3 | 2 | 1352 | |||
| GenBank mRNA | AK030506.1 | - | 1334 | |||
| AK050507.1 | - | 2 | 1334 | |||
| AK077649.1 | - | 2 | 1334 | |||
| AK167401.1 | - | 2 | 1353 | |||
| AK169400.1 | - | 2 | 1285 | |||
| BC016098.1 | - | 1 | 1249 | 4910571 | ||
| BC034309.1 | - | 1 | 1108 | 5362794 | ||

AW209491
Genomic Map