siRNA Details
| Gene Symbol: Kctd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111028.1 | NP_001104498.1 | 7, 8 | 796 | ||
| NM_001285933.1 | NP_001272862.1 | 7, 8 | 923 | |||
| NM_134073.2 | NP_598834.2 | 7, 8 | 796 | |||
| GenBank mRNA | AK087312.1 | - | 7, 8 | 668 | ||
| AK158922.1 | - | 7, 8 | 798 | |||
| AK166501.1 | - | 7, 8 | 632 | |||
| BC011302.1 | - | 7, 8 | 737 | |||
| CK032521.1 | - | 6, 7 | 433 | |||

Kctd9
Genomic Map