siRNA Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134084.1 | NP_598845.1 | 6 | 569 | ||
| GenBank mRNA | AK028349.1 | - | 4 | 2662 | ||
| BC004041.1 | - | 6 | 569 | 3496285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppif| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134084.1 | NP_598845.1 | 6 | 569 | ||
| GenBank mRNA | AK028349.1 | - | 4 | 2662 | ||
| BC004041.1 | - | 6 | 569 | 3496285 | ||