siRNA Details
| Gene Symbol: Slc38a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166456.1 | NP_001159928.1 | 12 | 1245 | ||
| NM_001166458.1 | NP_001159930.1 | 12 | 1452 | |||
| NM_134086.4 | NP_598847.2 | 12 | 1938 | |||
| GenBank mRNA | AF184240.2 | - | 11 | 1030 | ||
| AK029189.1 | - | 12 | 1115 | |||
| AK034130.1 | - | 12 | 1940 | |||
| AK035098.1 | - | 11 | 957 | |||
| AK047459.1 | - | 12 | 1469 | |||
| AK049173.1 | - | 12 | 1153 | |||
| AK049294.1 | - | 12 | 1218 | |||
| AK050914.1 | - | 12 | 1153 | |||
| AK081724.1 | - | 12 | 1454 | |||
| AK082683.1 | - | 12 | 1156 | |||
| AK162612.1 | - | 12 | 1303 | |||
| AK163914.1 | - | 7 | 556 | |||
| AK165188.1 | - | 12 | 1505 | |||
| BC030378.1 | - | 12 | 1154 | 5375319 | ||

Slc38a1
Genomic Map