siRNA Details
| Gene Symbol: Mal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178920.4 | NP_849251.1 | 3 | 551 | ||
| GenBank mRNA | AK035986.1 | - | 3 | 433 | ||
| BC113154.1 | - | 3 | 428 | 40058489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mal2| Gene Symbol: Mal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178920.4 | NP_849251.1 | 3 | 551 | ||
| GenBank mRNA | AK035986.1 | - | 3 | 433 | ||
| BC113154.1 | - | 3 | 428 | 40058489 | ||