siRNA Details
| Gene Symbol: Mfsd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134100.4 | NP_598861.1 | 2 | 971 | ||
| GenBank mRNA | AK087550.1 | - | 2 | 976 | ||
| AK089268.1 | - | 2 | 974 | |||
| AK165774.1 | - | 2 | 946 | |||
| AK194400.1 | - | 1 | 39 | |||
| BC009140.1 | - | 1 | 878 | 3598007 | ||

Mfsd5
Genomic Map