siRNA Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178925.3 | NP_849256.1 | 3 | 212 | ||
| GenBank mRNA | AK032127.1 | - | 3 | 254 | ||
| AK084698.1 | - | 3 | 206 | |||
| BC125628.1 | - | 3 | 212 | 40130143 | ||
| BC125630.1 | - | 3 | 212 | 40130145 | ||

Nsun3
Genomic Map