siRNA Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178925.3 | NP_849256.1 | 4, 5 | 665 | ||
| GenBank mRNA | AK032127.1 | - | 4, 5 | 707 | ||
| AK084698.1 | - | 4, 5 | 659 | |||
| AK153942.1 | - | 4, 5 | 479 | |||
| AK169841.1 | - | 4, 5 | 468 | |||
| BC125628.1 | - | 4, 5 | 665 | 40130143 | ||
| BC125630.1 | - | 4, 5 | 665 | 40130145 | ||

Nsun3
Genomic Map