siRNA Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134115.2 | NP_598876.1 | 6 | 638 | ||
| GenBank mRNA | AK035731.1 | - | 6 | 647 | ||
| AY292399.1 | - | 6 | 502 | |||
| BC009658.1 | - | 6 | 638 | 3484764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk38| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134115.2 | NP_598876.1 | 6 | 638 | ||
| GenBank mRNA | AK035731.1 | - | 6 | 647 | ||
| AY292399.1 | - | 6 | 502 | |||
| BC009658.1 | - | 6 | 638 | 3484764 | ||