siRNA Details
| Gene Symbol: Dlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286013.1 | NP_001272942.1 | 6 | 730 | ||
| NM_001286028.1 | NP_001272957.1 | 6 | 617 | |||
| GenBank mRNA | BC118057.1 | - | 6 | 617 | 40105170 | |
| BC122518.1 | - | 7 | 645 | 40105175 | ||
| BF472059.1 | - | 1 | 494 | |||
| BM948359.1 | - | 2 | 242 | |||
| FM180474.1 | - | 6 | 730 | |||

Dlk2
Genomic Map