siRNA Details
| Gene Symbol: Ankrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025572.1 | NP_001020743.1 | 9 | 4281 | ||
| GenBank mRNA | AK122392.1 | - | 1 | 642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd12| Gene Symbol: Ankrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025572.1 | NP_001020743.1 | 9 | 4281 | ||
| GenBank mRNA | AK122392.1 | - | 1 | 642 | ||