siRNA Details
| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 17 | 2942 | ||
| GenBank mRNA | AK129167.1 | - | 14 | 1633 | ||
| AK149331.1 | - | 17 | 2171 | |||
| AK163782.1 | - | 17 | 2570 | |||
| BC139005.1 | - | 17 | 2171 | 8862027 | ||
| BC145494.1 | - | 17 | 2172 | |||
| DQ266093.1 | - | 17 | 2305 | |||

Ift140
Genomic Map