siRNA Details
| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 21 | 3690 | ||
| GenBank mRNA | AK129167.1 | - | 18 | 2381 | ||
| AK149331.1 | - | 22 | 2955 | |||
| AK163782.1 | - | 21 | 3318 | |||
| AK172033.1 | - | 6 | 826 | |||
| BC009019.1 | - | 5 | 544 | |||
| BC139005.1 | - | 21 | 2919 | 8862027 | ||
| BC145494.1 | - | 21 | 2920 | |||
| DQ266093.1 | - | 21 | 3053 | |||

Ift140
Genomic Map