siRNA Details
| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 23, 24 | 4005 | ||
| GenBank mRNA | AK149331.1 | - | 24, 25 | 3270 | ||
| AK163782.1 | - | 23, 24 | 3633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift140| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 23, 24 | 4005 | ||
| GenBank mRNA | AK149331.1 | - | 24, 25 | 3270 | ||
| AK163782.1 | - | 23, 24 | 3633 | |||