siRNA Details
| Gene Symbol: Abhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134130.1 | NP_598891.1 | 6 | 779 | ||
| GenBank mRNA | AF429302.1 | - | 6 | 779 | ||
| AK134676.1 | - | 6 | 795 | |||
| AK144238.1 | - | 6 | 799 | |||
| BC026770.1 | - | 6 | 779 | 4234600 | ||

Abhd3
Genomic Map