siRNA Details
| Gene Symbol: Slc39a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139143.3 | NP_631882.2 | 7 | 2147 | ||
| GenBank mRNA | AB071697.1 | - | 6 | 1530 | ||
| AK028976.1 | - | 7 | 2106 | |||
| BC054780.1 | - | 6 | 1060 | 6813896 | ||
| BC055012.1 | - | 7 | 1817 | 6337369 | ||

Slc39a6
Genomic Map