siRNA Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177360.1 | NP_001170831.1 | 1 | 663 | ||
| NM_207220.2 | NP_997103.1 | 1 | 663 | |||
| GenBank mRNA | AK039612.1 | - | 1 | 163 | ||
| AK142928.1 | - | 1 | 261 | |||
| BC048078.1 | - | 2 | 535 | 6308873 | ||
| CX200961.1 | - | 1 | 663 | |||

Gpr137
Genomic Map