siRNA Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177360.1 | NP_001170831.1 | 2 | 821 | ||
| NM_001177361.1 | NP_001170832.1 | 4 | 994 | |||
| NM_001177362.1 | NP_001170833.1 | 4 | 685 | |||
| NM_207220.2 | NP_997103.1 | 4 | 1097 | |||
| GenBank mRNA | AK039612.1 | - | 4 | 597 | ||
| AK142928.1 | - | 2 | 419 | |||
| AK143450.1 | - | 4 | 995 | |||
| AK170308.1 | - | 4 | 687 | |||
| BC048078.1 | - | 5 | 969 | 6308873 | ||

Gpr137
Genomic Map