siRNA Details
| Gene Symbol: Ffar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181748.2 | NP_861413.1 | 3 | 1111 | ||
| GenBank mRNA | AB115769.1 | - | 3 | 1063 | ||
| AK131616.1 | - | 3 | 1098 | |||
| AY288424.1 | - | 3 | 1063 | |||
| BC053698.1 | - | 3 | 1111 | 6529120 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ffar4
Genomic Map