siRNA Details
| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA467412.1 | - | 3 | 319 | ||
| AK153119.1 | - | 3 | 232 | |||
| BE851841.1 | - | 1 | 411 | |||
| EF660528.1 | - | 3 | 336 | |||
| EH107460.1 | - | 3 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW112010| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA467412.1 | - | 3 | 319 | ||
| AK153119.1 | - | 3 | 232 | |||
| BE851841.1 | - | 1 | 411 | |||
| EF660528.1 | - | 3 | 336 | |||
| EH107460.1 | - | 3 | 269 | |||