siRNA Details
| Gene Symbol: Atf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030693.2 | NP_109618.1 | 3 | 781 | ||
| GenBank mRNA | AF310624.1 | - | 3 | 802 | ||
| AF312938.1 | - | 3 | 757 | |||
| AF375475.1 | - | 3 | 786 | |||
| AF375476.1 | - | 3 | 781 | |||
| AK131939.1 | - | 3 | 803 | |||
| AK146479.1 | - | 3 | 803 | |||
| AK159624.1 | - | 3 | 805 | |||
| AK160085.1 | - | 1 | 206 | |||
| AY024362.1 | - | 1 | 296 | |||
| BC010195.1 | - | 2 | 614 | 3157499 | ||
| BC092068.1 | - | 3 | 786 | 4210931 | ||

Atf5
Genomic Map