siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139293.2 | NP_647454.2 | 9 | 1248 | ||
| NM_177940.1 | NP_808809.1 | 9 | 1468 | |||
| NM_177941.1 | NP_808810.1 | 10 | 1555 | |||
| NM_177942.1 | NP_808811.1 | 8 | 1161 | |||
| GenBank mRNA | AF396699.1 | - | 9 | 1189 | ||
| AK031951.1 | - | 9 | 1256 | |||
| AK043473.1 | - | 7 | 911 | |||
| BC030900.1 | - | 4 | 371 | |||
| BC138052.1 | - | 9 | 1155 | 8861071 | ||
| BC138053.1 | - | 9 | 1155 | 8861072 | ||
| BC145126.1 | - | 9 | 1155 | |||

Ece2
Genomic Map