siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139293.2 | NP_647454.2 | 13 | 1683 | ||
| NM_177940.1 | NP_808809.1 | 13 | 1903 | |||
| NM_177941.1 | NP_808810.1 | 14 | 1990 | |||
| NM_177942.1 | NP_808811.1 | 12 | 1596 | |||
| GenBank mRNA | AF396699.1 | - | 13 | 1624 | ||
| AF489569.1 | - | 13 | 1722 | |||
| AF489570.1 | - | 14 | 1809 | |||
| AF489571.1 | - | 12 | 1596 | |||
| AF489572.1 | - | 13 | 1683 | |||
| AK031951.1 | - | 13 | 1691 | |||
| AK043473.1 | - | 11 | 1346 | |||
| AK134788.1 | - | 4 | 369 | |||
| BC030900.1 | - | 8 | 806 | |||
| BC138052.1 | - | 13 | 1590 | 8861071 | ||
| BC138053.1 | - | 13 | 1590 | 8861072 | ||
| BC145126.1 | - | 13 | 1590 | |||

Ece2
Genomic Map