siRNA Details
| Gene Symbol: Mylk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139300.3 | NP_647461.3 | 16 | 2832 | ||
| GenBank mRNA | AK044527.1 | - | 6 | 1068 | ||
| AK078556.1 | - | 1 | 271 | |||
| AY237726.1 | - | 16 | 2861 | |||
| AY237727.2 | - | 1 | 385 | |||
| BC030478.1 | - | 6 | 1064 | |||
| BC045197.1 | - | 1 | 315 | |||
| BC058610.1 | - | 16 | 2844 | 30056142 | ||
| BC145279.1 | - | 16 | 2819 | 9053833 | ||
| BC150749.1 | - | 16 | 2819 | 9087660 | ||
| BC171954.1 | - | 16 | 2820 | |||

Mylk
Genomic Map