siRNA Details
| Gene Symbol: Rdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080436.3 | NP_536684.2 | 3 | 865 | ||
| GenBank mRNA | AK136574.1 | - | 3 | 908 | ||
| AY028928.1 | - | 3 | 819 | |||
| BC137930.1 | - | 3 | 783 | 8860949 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh1| Gene Symbol: Rdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080436.3 | NP_536684.2 | 3 | 865 | ||
| GenBank mRNA | AK136574.1 | - | 3 | 908 | ||
| AY028928.1 | - | 3 | 819 | |||
| BC137930.1 | - | 3 | 783 | 8860949 | ||