siRNA Details
| Gene Symbol: Aldh1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027406.1 | NP_081682.1 | 11 | 1423 | ||
| GenBank mRNA | BC022112.1 | - | 10 | 1376 | ||
| BC024055.1 | - | 11 | 1380 | 5098789 | ||
| BC025939.1 | - | 11 | 1405 | 5099360 | ||
| BC028817.1 | - | 11 | 1370 | 5101815 | ||
| BC030722.1 | - | 11 | 1407 | 5098671 | ||
| BC030723.1 | - | 11 | 1339 | 5100007 | ||
| BC030727.1 | - | 11 | 1409 | 5099026 | ||
| BC030730.1 | - | 11 | 1423 | 5101835 | ||

Aldh1l1
Genomic Map