siRNA Details
| Gene Symbol: Gtpbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145147.5 | NP_660129.2 | 4 | 602 | ||
| GenBank mRNA | AJ293619.1 | - | 3 | 232 | ||
| AK082192.1 | - | 1 | 478 | |||
| AK144812.1 | - | 4 | 603 | |||
| AK152225.1 | - | 4 | 603 | |||
| AK152972.1 | - | 4 | 603 | |||
| AK159462.1 | - | 4 | 575 | |||
| BC027821.1 | - | 1 | 133 | 5321725 | ||

Gtpbp6
Genomic Map