siRNA Details
| Gene Symbol: Lin7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033223.2 | NP_001028395.1 | 5 | 534 | ||
| NM_001039354.1 | NP_001034443.1 | 5 | 576 | |||
| NM_001284329.1 | NP_001271258.1 | 5 | 1293 | |||
| GenBank mRNA | AK077191.1 | - | 5 | 1293 | ||
| AK160671.1 | - | 5 | 483 | |||
| BC029721.1 | - | 5 | 482 | |||
| BG288381.1 | - | 5 | 514 | |||
| BU923246.1 | - | 5 | 543 | |||

Lin7a
Genomic Map