siRNA Details
| Gene Symbol: Eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201256.4 | NP_957708.1 | 1, 2 | 250 | ||
| GenBank mRNA | AY226182.1 | - | 1, 2 | 128 | ||
| BC038258.1 | - | 1, 2 | 157 | |||
| BC061242.1 | - | 1, 2 | 250 | 6743910 | ||
| BC111107.1 | - | 1, 2 | 190 | 40055518 | ||

Eif4ebp3
Genomic Map