siRNA Details
| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081020.1 | NP_001074489.1 | 3 | 830 | ||
| GenBank mRNA | AK030315.1 | - | 3 | 982 | ||
| AK044528.1 | - | 3 | 833 | |||
| BC172116.1 | - | 3 | 728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts6| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081020.1 | NP_001074489.1 | 3 | 830 | ||
| GenBank mRNA | AK030315.1 | - | 3 | 982 | ||
| AK044528.1 | - | 3 | 833 | |||
| BC172116.1 | - | 3 | 728 | |||