siRNA Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138607.3 | NP_613073.2 | 1, 2 | 289 | ||
| GenBank mRNA | AK008402.1 | - | 1, 2 | 290 | ||
| BC026128.1 | - | 1, 2 | 199 | |||
| Y18505.1 | - | 1, 2 | 254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam50a| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138607.3 | NP_613073.2 | 1, 2 | 289 | ||
| GenBank mRNA | AK008402.1 | - | 1, 2 | 290 | ||
| BC026128.1 | - | 1, 2 | 199 | |||
| Y18505.1 | - | 1, 2 | 254 | |||