siRNA Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138607.3 | NP_613073.2 | 9 | 938 | ||
| GenBank mRNA | AK008402.1 | - | 9 | 939 | ||
| BC026128.1 | - | 8 | 1311 | |||
| BC125297.1 | - | 8 | 804 | 40129812 | ||
| Y18505.1 | - | 9 | 903 | |||

Fam50a
Genomic Map