siRNA Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199274.1 | NP_001186203.1 | 6, 7 | 1164 | ||
| NM_134017.2 | NP_598778.1 | 6, 7 | 968 | |||
| GenBank mRNA | AB070267.1 | - | 6, 7 | 935 | ||
| AK075600.1 | - | 6, 7 | 869 | |||
| AK075947.1 | - | 6, 7 | 903 | |||
| AK153126.1 | - | 6, 7 | 922 | |||
| AK162087.1 | - | 6, 7 | 903 | |||
| BC003457.1 | - | 6, 7 | 899 | 2648452 | ||

Mat2b
Genomic Map