siRNA Details
| Gene Symbol: Rnf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022423.2 | NP_071868.2 | 2 | 576 | ||
| GenBank mRNA | AB030190.1 | - | 2 | 568 | ||
| AK037991.1 | - | 2 | 491 | |||
| AK075944.1 | - | 2 | 587 | |||
| AK076158.1 | - | 2 | 566 | |||
| AK088973.1 | - | 2 | 579 | |||
| BC072613.1 | - | 2 | 569 | |||
| BC096522.1 | - | 2 | 540 | |||

Rnf187
Genomic Map