siRNA Details
| Gene Symbol: Rnf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022423.2 | NP_071868.2 | 3 | 815 | ||
| GenBank mRNA | AB030190.1 | - | 3 | 807 | ||
| AK037991.1 | - | 3 | 730 | |||
| AK075944.1 | - | 3 | 826 | |||
| AK076158.1 | - | 3 | 805 | |||
| AK088973.1 | - | 3 | 818 | |||
| AK213464.1 | - | 1 | 15 | |||
| BC072613.1 | - | 3 | 808 | |||
| BC096522.1 | - | 3 | 779 | |||

Rnf187
Genomic Map