siRNA Details
| Gene Symbol: Mex3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175366.3 | NP_780575.2 | 2 | 1331 | ||
| GenBank mRNA | AK173330.1 | - | 1 | 2144 | ||
| BC031512.1 | - | 1 | 315 | |||
| BC067199.1 | - | 1 | 296 | |||
| BC108401.1 | - | 1 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mex3b| Gene Symbol: Mex3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175366.3 | NP_780575.2 | 2 | 1331 | ||
| GenBank mRNA | AK173330.1 | - | 1 | 2144 | ||
| BC031512.1 | - | 1 | 315 | |||
| BC067199.1 | - | 1 | 296 | |||
| BC108401.1 | - | 1 | 539 | |||