siRNA Details
| Gene Symbol: Aif1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145144.1 | NP_660126.1 | 2 | 120 | ||
| GenBank mRNA | AB035322.1 | - | 2 | 72 | ||
| AK028955.1 | - | 2 | 78 | |||
| AK045539.1 | - | 2 | 150 | |||
| BC024599.1 | - | 2 | 120 | 5044495 | ||

Aif1l
Genomic Map