siRNA Details
| Gene Symbol: 2700081O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175381.6 | NP_780590.3 | 4 | 1209 | ||
| GenBank mRNA | BC059063.1 | - | 2 | 648 | 6825195 | |
| BC096549.1 | - | 3 | 248 | |||
| BC096638.1 | - | 2 | 434 | 6825382 | ||

2700081O15Rik
Genomic Map