siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242407.1 | NP_001229336.1 | 13 | 1598 | ||
| NM_175392.3 | NP_780601.1 | 13 | 1766 | |||
| GenBank mRNA | AK077618.1 | - | 13 | 1559 | ||
| AK131152.1 | - | 10 | 2448 | |||
| AK154200.1 | - | 13 | 1587 | |||
| AK165996.1 | - | 13 | 1590 | |||
| BC051404.1 | - | 13 | 1766 | 6395030 | ||

Fam73b
Genomic Map