siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242407.1 | NP_001229336.1 | 2 | 281 | ||
| NM_175392.3 | NP_780601.1 | 2 | 449 | |||
| GenBank mRNA | AK040713.1 | - | 2 | 250 | ||
| AK077618.1 | - | 2 | 242 | |||
| AK140122.1 | - | 2 | 267 | |||
| AK154200.1 | - | 2 | 270 | |||
| AK165996.1 | - | 2 | 272 | |||
| BC051404.1 | - | 2 | 449 | 6395030 | ||

Fam73b
Genomic Map