siRNA Details
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175401.3 | NP_780610.2 | 9, 10 | 1303 | ||
GenBank mRNA | AK075676.1 | - | 1, 2 | 1428 | ||
AK135553.1 | - | 7, 8 | 2155 | |||
AK155157.1 | - | 10, 11 | 1817 | |||
BC040428.1 | - | 9, 10 | 1303 | 3327345 |
