siRNA Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175403.3 | NP_780612.2 | 1, 2 | 380 | ||
| GenBank mRNA | AK031525.1 | - | 1, 2 | 350 | ||
| AK075940.1 | - | 1, 2 | 364 | |||
| AK129066.1 | - | 1, 2 | 283 | |||
| BC035300.1 | - | 1, 2 | 345 | 4237864 | ||
| BC058245.1 | - | 1, 2 | 334 | 6313259 | ||
| CJ093242.1 | - | 1, 2 | 380 | |||

Mlec
Genomic Map