siRNA Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175403.3 | NP_780612.2 | 3 | 639 | ||
| GenBank mRNA | AK031525.1 | - | 3 | 609 | ||
| AK075940.1 | - | 3 | 623 | |||
| AK129066.1 | - | 3 | 542 | |||
| BC035300.1 | - | 3 | 604 | 4237864 | ||
| BC058245.1 | - | 3 | 593 | 6313259 | ||

Mlec
Genomic Map