siRNA Details
| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175408.4 | NP_780617.2 | 3 | 582 | ||
| GenBank mRNA | AK053212.1 | - | 1 | 2033 | ||
| AK080737.1 | - | 1 | 805 | |||
| BC096053.1 | - | 1 | 265 | 3467410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem139| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175408.4 | NP_780617.2 | 3 | 582 | ||
| GenBank mRNA | AK053212.1 | - | 1 | 2033 | ||
| AK080737.1 | - | 1 | 805 | |||
| BC096053.1 | - | 1 | 265 | 3467410 | ||