siRNA Details
Gene Symbol: Tmem139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175408.4 | NP_780617.2 | 3 | 705 | ||
GenBank mRNA | AK053212.1 | - | 1 | 2156 | ||
AK080737.1 | - | 1 | 682 | |||
BC096053.1 | - | 1 | 388 | 3467410 |
