siRNA Details
| Gene Symbol: Kif24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024241.2 | NP_077203.2 | 11 | 3909 | ||
| GenBank mRNA | AK084967.1 | - | 8 | 3508 | ||
| BC067395.1 | - | 11 | 3901 | 30460089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif24| Gene Symbol: Kif24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024241.2 | NP_077203.2 | 11 | 3909 | ||
| GenBank mRNA | AK084967.1 | - | 8 | 3508 | ||
| BC067395.1 | - | 11 | 3901 | 30460089 | ||