siRNA Details
| Gene Symbol: Ssc4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160366.1 | NP_001153838.1 | 10 | 1674 | ||
| GenBank mRNA | AK082786.1 | - | 2 | 85 | ||
| BC117747.1 | - | 7 | 1181 | |||
| BC126947.1 | - | 7 | 1181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssc4d| Gene Symbol: Ssc4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160366.1 | NP_001153838.1 | 10 | 1674 | ||
| GenBank mRNA | AK082786.1 | - | 2 | 85 | ||
| BC117747.1 | - | 7 | 1181 | |||
| BC126947.1 | - | 7 | 1181 | |||