siRNA Details
| Gene Symbol: R3hdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177994.4 | NP_818775.2 | 2 | 172 | ||
| GenBank mRNA | AK081288.1 | - | 2 | 161 | ||
| AK088107.1 | - | 2 | 173 | |||
| AK139047.1 | - | 2 | 163 | |||
| AK159869.1 | - | 2 | 151 | |||
| AK165044.1 | - | 1 | 3281 | |||
| AK165610.1 | - | 2 | 143 | |||
| BC095970.1 | - | 2 | 153 | 6530790 | ||
| BC132463.1 | - | 2 | 248 | 40130740 | ||
| BC138461.1 | - | 2 | 248 | 8861481 | ||

R3hdm4
Genomic Map